missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Hydroxypyridazine-3-carboxylic Acid Monohydrate 98.0+%, TCI America™
$56.41 - $169.22
Chemical Identifiers
CAS | 306934-80-5 |
---|---|
Molecular Formula | C5H6N2O4 |
Molecular Weight (g/mol) | 158.113 |
MDL Number | MFCD09064936 |
InChI Key | DZCBVXKRERSHAA-UHFFFAOYSA-N |
PubChem CID | 2774651 |
IUPAC Name | 6-oxo-1H-pyridazine-3-carboxylic acid;hydrate |
SMILES | C1=CC(=O)NN=C1C(=O)O.O |
Chemical Identifiers
306934-80-5 | |
158.113 | |
DZCBVXKRERSHAA-UHFFFAOYSA-N | |
6-oxo-1H-pyridazine-3-carboxylic acid;hydrate |
C5H6N2O4 | |
MFCD09064936 | |
2774651 | |
C1=CC(=O)NN=C1C(=O)O.O |
Specifications
306934-80-5 | |
White | |
MFCD09064936 | |
DZCBVXKRERSHAA-UHFFFAOYSA-N | |
6-oxo-1H-pyridazine-3-carboxylic acid;hydrate | |
2774651 | |
≥98.0% (HPLC,T) | |
6-Hydroxypyridazine-3-carboxylic Acid Monohydrate |
257°C | |
C5H6N2O4 | |
1 g | |
C1=CC(=O)NN=C1C(=O)O.O | |
158.113 | |
140.10 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-0974
TSCA : No