missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Hydroxynaphthalene-2-boronic acid pinacol ester, 96%, Thermo Scientific™
$262.58 - $778.97
Chemical Identifiers
CAS | 269410-21-1 |
---|---|
Molecular Formula | C16H19BO3 |
Molecular Weight (g/mol) | 270.135 |
MDL Number | MFCD20486767 |
InChI Key | FEOQRVDXBOSJLZ-UHFFFAOYSA-N |
Synonym | 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl naphthalen-2-ol, 6-hydroxynaphthalene-2-boronic acid pinacol ester, 6-hydroxy-2-naphthaleneboronic acid, pinacol ester, 6-tetramethyl-1,3,2-dioxaborolan-2-yl naphthalen-2-ol, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl naphthalen-2-ol |
PubChem CID | 21923963 |
IUPAC Name | 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C=C(C=C3)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH62631MD
|
Thermo Scientific Chemicals
H62631MD |
250 mg |
Each for $262.58
|
|
|||||
AAH6263103
|
Thermo Scientific Chemicals
H6263103 |
1 g |
Each for $778.97
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
269410-21-1 | |
270.135 | |
FEOQRVDXBOSJLZ-UHFFFAOYSA-N | |
21923963 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C=C(C=C3)O |
C16H19BO3 | |
MFCD20486767 | |
6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl naphthalen-2-ol, 6-hydroxynaphthalene-2-boronic acid pinacol ester, 6-hydroxy-2-naphthaleneboronic acid, pinacol ester, 6-tetramethyl-1,3,2-dioxaborolan-2-yl naphthalen-2-ol, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl naphthalen-2-ol | |
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol |
Specifications
269410-21-1 | |
C16H19BO3 | |
250 mg | |
FEOQRVDXBOSJLZ-UHFFFAOYSA-N | |
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol | |
21923963 | |
96% | |
6-Hydroxynaphthalene-2-boronic acid pinacol ester |
White | |
MFCD20486767 | |
6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl naphthalen-2-ol, 6-hydroxynaphthalene-2-boronic acid pinacol ester, 6-hydroxy-2-naphthaleneboronic acid, pinacol ester, 6-tetramethyl-1,3,2-dioxaborolan-2-yl naphthalen-2-ol, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl naphthalen-2-ol | |
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C=C(C=C3)O | |
270.135 | |
270.13 | |
Solid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures