Learn More
6-Hydroxyindole, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5595103
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
6-Hydroxyindole | |
125°C to 128°C | |
MFCD00152101 | |
UN3077 | |
XAWPKHNOFIWWNZ-UHFFFAOYSA-N | |
1H-indol-6-ol | |
524508 | |
98% |
2380-86-1 | |
C8H7NO | |
1 g | |
6-hydroxyindole, 6-indolol, indolol, 6-hydroxy indole, unii-3i03jz599t, 6-hydroxy-1h-indole, 6hydroxyindole, 6-hydroxy-indole, indol-6-ol, 1h-indole-6-ol | |
C1=CC(=CC2=C1C=CN2)O | |
133.15 | |
133.15 |
Chemical Identifiers
2380-86-1 | |
133.15 | |
XAWPKHNOFIWWNZ-UHFFFAOYSA-N | |
524508 | |
C1=CC(=CC2=C1C=CN2)O |
C8H7NO | |
MFCD00152101 | |
6-hydroxyindole, 6-indolol, indolol, 6-hydroxy indole, unii-3i03jz599t, 6-hydroxy-1h-indole, 6hydroxyindole, 6-hydroxy-indole, indol-6-ol, 1h-indole-6-ol | |
1H-indol-6-ol |
Safety and Handling
GHS H Statement
H318-H302-H317
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P501c
H302-H317-H318
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 417-020-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only