Please login to your online account to display your discounted pricing

6-Hydroxyindole, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC379010250

 View more versions of this product

Catalog No. AC379010250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 6-Hydroxyindole
Color White to off-white
Melting Point 124° to 130°C
Quantity 25g
CAS 2380-86-1
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Shiny Crystalline Powder
Assay ≥97.5% (HPLC)
Assay Percent Range 97%
Molecular Formula C8H7NO
Storage Information Product may darken during storage
Formula Weight 133.15

Specifications

Chemical Name or Material 6-Hydroxyindole
Assay ≥97.5% (HPLC)
Assay Percent Range 97%
Molecular Formula C8H7NO
Formula Weight 133.15