missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Hydroxyhexanoic acid, 95%, may cont. variable amounts of dimer, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2485703

 View more versions of this product

Catalog No. AAB2485703



Description

Description

6-Hydroxyhexanoic acid acts as a useful reagent in synthetic chemistry. It is used as a precursor to prepare surface active monomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Hydroxyhexanoic acid acts as a useful reagent in synthetic chemistry. It is used as a precursor to prepare surface active monomers.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

6-Hydroxyhexanoic acid
38°C to 40°C
>110°C (230°F)
C6H12O3
HO(CH2)5CO2H
1 g
IWHLYPDWHHPVAA-UHFFFAOYSA-N
6-hydroxyhexanoic acid
14490
132.16
1191-25-9
1.08
may cont. variable amounts of dimer
1.456
MFCD00046560
6-hydroxycaproic acid, 6-hydroxyhexanoate, hexanoic acid, 6-hydroxy, 6-hydroxy caproic acid, 6-hydroxy-hexanoic acid, epsilon-hydroxycaproic acid, 5-carboxypentanol, unii-3y3ox37nm8, 6-hydroxycaproicacid,pract.,containslactone, 6-hydroxy-caproic acid
C(CCC(=O)O)CCO
132.159
CHEBI:17869
95%

Chemical Identifiers

1191-25-9
132.159
IWHLYPDWHHPVAA-UHFFFAOYSA-N
14490
6-hydroxyhexanoic acid
C6H12O3
MFCD00046560
6-hydroxycaproic acid, 6-hydroxyhexanoate, hexanoic acid, 6-hydroxy, 6-hydroxy caproic acid, 6-hydroxy-hexanoic acid, epsilon-hydroxycaproic acid, 5-carboxypentanol, unii-3y3ox37nm8, 6-hydroxycaproicacid,pract.,containslactone, 6-hydroxy-caproic acid
CHEBI:17869
C(CCC(=O)O)CCO
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only