missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Hydroxyflavanone, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2375503

 View more versions of this product

Catalog No. AAB2375503



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

6-Hydroxyflavanone
∼220°C (decomposition)
MFCD00017485
87354
XYHWPQUEOOBIOW-UHFFFAOYSA-N
6-hydroxy-2-phenyl-2,3-dihydrochromen-4-one
2734580
240.25
4250-77-5
C15H12O3
1 g
6-hydroxyflavanone, 6-hydroxy-2-phenylchroman-4-one, 6-hydroxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 6-hydroxy-flavanone, 4h-1-benzopyran-4-one, 2,3-dihydro-6-hydroxy-2-phenyl, 6-hydroxy-2-phenyl-chroman-4-one, flavanone, 6-hydroxy, +/--6-hydroxyflavanone, acmc-209jpu
C1C(OC2=C(C1=O)C=C(C=C2)O)C3=CC=CC=C3
240.258
CHEBI:34471
≥98%

Chemical Identifiers

4250-77-5
240.258
XYHWPQUEOOBIOW-UHFFFAOYSA-N
2734580
6-hydroxy-2-phenyl-2,3-dihydrochromen-4-one
C15H12O3
MFCD00017485
6-hydroxyflavanone, 6-hydroxy-2-phenylchroman-4-one, 6-hydroxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 6-hydroxy-flavanone, 4h-1-benzopyran-4-one, 2,3-dihydro-6-hydroxy-2-phenyl, 6-hydroxy-2-phenyl-chroman-4-one, flavanone, 6-hydroxy, +/--6-hydroxyflavanone, acmc-209jpu
CHEBI:34471
C1C(OC2=C(C1=O)C=C(C=C2)O)C3=CC=CC=C3
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only