missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Hydroxycoumarin, TCI America™

$151.25 - $462.92

Chemical Identifiers

CAS 6093-68-1
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.144
MDL Number MFCD00630040
InChI Key CJIJXIFQYOPWTF-UHFFFAOYSA-N
Synonym 6-hydroxycoumarin, 6-hydroxy-2h-chromen-2-one, 2h-1-benzopyran-2-one, 6-hydroxy, 6-hydroxycoumaran, 2h-1-benzopyran-2-one,6-hydroxy, 6-hydroxbenzopyran-2-one, 6-hydroxy-2h-chromen-2-on, 6-hydroxycoumarine, 6-hydroxy coumarin, 6-hydroxy-2h-1-benzopyran-2-one
PubChem CID 99477
IUPAC Name 6-hydroxychromen-2-one
SMILES C1=CC2=C(C=CC(=O)O2)C=C1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H13275G View Documents TCI America
H13275G
5g Each for $151.25
 
H132725G View Documents TCI America
H132725G
25g Each for $462.92
 
Specifications

Chemical Identifiers

6093-68-1
162.144
CJIJXIFQYOPWTF-UHFFFAOYSA-N
99477
C1=CC2=C(C=CC(=O)O2)C=C1O
C9H6O3
MFCD00630040
6-hydroxycoumarin, 6-hydroxy-2h-chromen-2-one, 2h-1-benzopyran-2-one, 6-hydroxy, 6-hydroxycoumaran, 2h-1-benzopyran-2-one,6-hydroxy, 6-hydroxbenzopyran-2-one, 6-hydroxy-2h-chromen-2-on, 6-hydroxycoumarine, 6-hydroxy coumarin, 6-hydroxy-2h-1-benzopyran-2-one
6-hydroxychromen-2-one

Specifications

Yellow
5g
C9H6O3
6-hydroxycoumarin, 6-hydroxy-2h-chromen-2-one, 2h-1-benzopyran-2-one, 6-hydroxy, 6-hydroxycoumaran, 2h-1-benzopyran-2-one,6-hydroxy, 6-hydroxbenzopyran-2-one, 6-hydroxy-2h-chromen-2-on, 6-hydroxycoumarine, 6-hydroxy coumarin, 6-hydroxy-2h-1-benzopyran-2-one
C1=CC2=C(C=CC(=O)O2)C=C1O
162.144
162.14
≥98.0% (GC,T)
255°C
6093-68-1
MFCD00630040
CJIJXIFQYOPWTF-UHFFFAOYSA-N
6-hydroxychromen-2-one
99477
Crystal-Powder at 20°C
6-Hydroxycoumarin
Safety and Handling

Safety and Handling

TSCA : No