missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Hydroxy-4-pyrimidinecarboxylic Acid, TCI America™

$53.47 - $160.42

Chemical Identifiers

CAS 6299-87-2
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.098
MDL Number MFCD00023267
InChI Key QYGHXDAYBIFGKI-UHFFFAOYSA-N
Synonym 1,6-Dihydro-6-oxo-4-pyrimidinecarboxylic Acid
PubChem CID 239692
IUPAC Name 4-oxo-1H-pyrimidine-6-carboxylic acid
SMILES C1=C(NC=NC1=O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H13921G View Documents TCI America
H13921G
1g Each for $160.42
 
H1392200MG View Documents TCI America
H1392200MG
200mg Each for $53.47
 
Specifications

Chemical Identifiers

6299-87-2
140.098
QYGHXDAYBIFGKI-UHFFFAOYSA-N
239692
C1=C(NC=NC1=O)C(=O)O
C5H4N2O3
MFCD00023267
1,6-Dihydro-6-oxo-4-pyrimidinecarboxylic Acid
4-oxo-1H-pyrimidine-6-carboxylic acid

Specifications

Undesignated
1g
C5H4N2O3
1,6-Dihydro-6-oxo-4-pyrimidinecarboxylic Acid
C1=C(NC=NC1=O)C(=O)O
140.098
140.10
≥98.0% (HPLC,T)
267°C
6299-87-2
MFCD00023267
QYGHXDAYBIFGKI-UHFFFAOYSA-N
4-oxo-1H-pyrimidine-6-carboxylic acid
239692
Crystal-Powder at 20°C
6-Hydroxy-4-pyrimidinecarboxylic Acid
Safety and Handling

Safety and Handling

TSCA : No