missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Hydroxy-4-methylcoumarin, TCI America™

$74.86 - $223.06

Chemical Identifiers

CAS 2373-31-1
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
MDL Number MFCD00016967
InChI Key IRUHWRSITUYICV-UHFFFAOYSA-N
Synonym 6-hydroxy-4-methylcoumarin, 6-hydroxy-4-methyl-2h-chromen-2-one, coumarin, 6-hydroxy-4-methyl, 6-hydroxy-4-methyl-2-benzopyrone, 6-hydroxy-4-methyl-chromen-2-one, 2h-1-benzopyran-2-one, 6-hydroxy-4-methyl, chembl12626, 2h-1-benzopyran-2-one,6-hydroxy-4-methyl, acmc-1cuho, 4-methyl-6-hydroxycoumarin
PubChem CID 75409
IUPAC Name 6-hydroxy-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=C1C=C(C=C2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H10055G View Documents TCI America
H10055G
5g Each for $74.86
 
H100525G View Documents TCI America
H100525G
25g Each for $223.06
 
Specifications

Chemical Identifiers

2373-31-1
176.171
IRUHWRSITUYICV-UHFFFAOYSA-N
75409
CC1=CC(=O)OC2=C1C=C(C=C2)O
C10H8O3
MFCD00016967
6-hydroxy-4-methylcoumarin, 6-hydroxy-4-methyl-2h-chromen-2-one, coumarin, 6-hydroxy-4-methyl, 6-hydroxy-4-methyl-2-benzopyrone, 6-hydroxy-4-methyl-chromen-2-one, 2h-1-benzopyran-2-one, 6-hydroxy-4-methyl, chembl12626, 2h-1-benzopyran-2-one,6-hydroxy-4-methyl, acmc-1cuho, 4-methyl-6-hydroxycoumarin
6-hydroxy-4-methylchromen-2-one

Specifications

Undesignated
5g
C10H8O3
6-hydroxy-4-methylcoumarin, 6-hydroxy-4-methyl-2h-chromen-2-one, coumarin, 6-hydroxy-4-methyl, 6-hydroxy-4-methyl-2-benzopyrone, 6-hydroxy-4-methyl-chromen-2-one, 2h-1-benzopyran-2-one, 6-hydroxy-4-methyl, chembl12626, 2h-1-benzopyran-2-one,6-hydroxy-4-methyl, acmc-1cuho, 4-methyl-6-hydroxycoumarin
CC1=CC(=O)OC2=C1C=C(C=C2)O
176.171
176.17
≥98.0% (GC,T)
248°C
2373-31-1
MFCD00016967
IRUHWRSITUYICV-UHFFFAOYSA-N
6-hydroxy-4-methylchromen-2-one
75409
Crystal-Powder at 20°C
6-Hydroxy-4-methylcoumarin
Safety and Handling

Safety and Handling

RTECSNumber : GN6999800

TSCA : No