Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Hydroxy-4-methylcoumarin, 99%

$26.93 - $26.93

Chemical Identifiers

Chemical Name or Material 6-Hydroxy-4-methylcoumarin
CAS 2373-31-1
Molecular Formula C10H8O3
Formula Weight 176.17
MDL Number MFCD00016967
Beilstein 144873
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0719203

  SDS
alfa aesar™
L0719203
1g Each for $26.93
Description & Specifications

Specifications

Formula Weight 176.17
Chemical Name or Material 6-Hydroxy-4-methylcoumarin
Assay 99%
Molecular Formula C10H8O3
MDL Number MFCD00016967
CAS 2373-31-1
Beilstein 144873
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 246° to 250°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.