Please login to your online account to display your discounted pricing

6-Hydroxy-3-nitro-2-picoline, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC360860050

 View more versions of this product

Catalog No. AC360860050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 154
Melting Point 230° to 232°C (subl.)
Physical Form Crystalline Powder
Quantity 5g
Color Light brown to brown
CAS Max % 100.0
Chemical Name or Material 6-Hydroxy-3-nitro-2-picoline
Assay Percent Range 98%
Molecular Formula C6H6N2O3
CAS 28489-45-4
Packaging Glass bottle
MDL Number MFCD00092010
CAS Min % 97.5
Health Hazard 2 GHS P Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 6-Hydroxy-3-nitro-2-picoline
CAS 28489-45-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H6N2O3
Formula Weight 154
MDL Number MFCD00092010
Accessories

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents