missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Hydroxy-3-nitro-2-picoline, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 28489-45-4 |
---|---|
Molecular Formula | C6H6N2O3 |
Molecular Weight (g/mol) | 154 |
MDL Number | MFCD00092010 |
InChI Key | AJDDHNXZBGZBJN-UHFFFAOYSA-N |
Synonym | 2 1h-pyridinone, 6-methyl-5-nitro, 2-hydroxy-5-nitro-6-picoline, 2-hydroxy-6-methyl-5-nitropyridine, 6-hydroxy-2-methyl-3-nitropyridine, 6-hydroxy-3-nitro-2-picoline, 6-methyl-5-nitro-2-pyridinol, 6-methyl-5-nitro-pyridin-2-ol, 6-methyl-5-nitropyridin-2 1h-one, 6-methyl-5-nitropyridin-2-ol, pubchem10387 |
PubChem CID | 543053 |
IUPAC Name | 6-methyl-5-nitro-1H-pyridin-2-one |
SMILES | CC1=C(C=CC(=O)N1)[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC360860010
|
Thermo Fisher Scientific
AC360860010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
28489-45-4 | |
154 | |
AJDDHNXZBGZBJN-UHFFFAOYSA-N | |
543053 | |
CC1=C(C=CC(=O)N1)[N+](=O)[O-] |
C6H6N2O3 | |
MFCD00092010 | |
2 1h-pyridinone, 6-methyl-5-nitro, 2-hydroxy-5-nitro-6-picoline, 2-hydroxy-6-methyl-5-nitropyridine, 6-hydroxy-2-methyl-3-nitropyridine, 6-hydroxy-3-nitro-2-picoline, 6-methyl-5-nitro-2-pyridinol, 6-methyl-5-nitro-pyridin-2-ol, 6-methyl-5-nitropyridin-2 1h-one, 6-methyl-5-nitropyridin-2-ol, pubchem10387 | |
6-methyl-5-nitro-1H-pyridin-2-one |
Specifications
28489-45-4 | |
100.0 | |
230°C to 232°C | |
Authentic | |
MFCD00092010 | |
2 1h-pyridinone, 6-methyl-5-nitro, 2-hydroxy-5-nitro-6-picoline, 2-hydroxy-6-methyl-5-nitropyridine, 6-hydroxy-2-methyl-3-nitropyridine, 6-hydroxy-3-nitro-2-picoline, 6-methyl-5-nitro-2-pyridinol, 6-methyl-5-nitro-pyridin-2-ol, 6-methyl-5-nitropyridin-2 1h-one, 6-methyl-5-nitropyridin-2-ol, pubchem10387 | |
AJDDHNXZBGZBJN-UHFFFAOYSA-N | |
6-methyl-5-nitro-1H-pyridin-2-one | |
543053 | |
Crystalline Powder | |
6-Hydroxy-3-nitro-2-picoline |
97.5 | |
Brown | |
97.5% min. (HPLC) | |
C6H6N2O3 | |
Glass bottle | |
1g | |
CC1=C(C=CC(=O)N1)[N+](=O)[O-] | |
154 | |
154 | |
98% |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
RUO – Research Use Only