Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Hydroxy-3-iodo-2-methylpyridine, 95%

$102.99 - $237.05

Chemical Identifiers

Chemical Name or Material 6-Hydroxy-3-iodo-2-methylpyridine
CAS 927870-76-6
Molecular Formula C6H6INO
Formula Weight 235.02
MDL Number MFCD09037468
Synonym 5-Iodo-6-methyl-2(1H)-pyridone; 3-Iodo-2-picolin-6-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27381MD

  SDS
alfa aesar™
H27381MD
250mg Each for $102.99

AAH2738103

  SDS
alfa aesar™
H2738103
1g Each for $237.05
Description & Specifications

Specifications

Formula Weight 235.02
Chemical Name or Material 6-Hydroxy-3-iodo-2-methylpyridine
Assay 95%
Molecular Formula C6H6INO
MDL Number MFCD09037468
CAS 927870-76-6
Synonym 5-Iodo-6-methyl-2(1H)-pyridone; 3-Iodo-2-picolin-6-ol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈152°C
Quantity 250mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.