missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Hydroxy-2H-chromene-3-carboxaldehyde, 97%

$86.05 - $246.28

Chemical Identifiers

CAS 134822-76-7
Molecular Formula C10H8O3
Formula Weight 176.17
MDL Number MFCD01076587
Beilstein 6646155
Synonym 6-Hydroxy-2H-1-benzopyran-3-carboxaldehyde
Chemical Name or Material 6-Hydroxy-2H-chromene-3-carboxaldehyde
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1474203 SDS Alfa Aesar™
L1474203
1g Each for $246.28
AAL14742MD SDS Alfa Aesar™
L14742MD
250mg Each for $86.05
Specifications Description & Specifications

Chemical Identifiers

134822-76-7
176.17
6646155
6-Hydroxy-2H-chromene-3-carboxaldehyde
C10H8O3
MFCD01076587
6-Hydroxy-2H-1-benzopyran-3-carboxaldehyde

Specifications

164° to 166°C
134822-76-7
176.17
6646155
6-Hydroxy-2H-chromene-3-carboxaldehyde
1g
C10H8O3
MFCD01076587
6-Hydroxy-2H-1-benzopyran-3-carboxaldehyde
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning