missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Hydroxy-2-pyridinecarboxylic Acid, TCI America™

$100.83 - $305.56

Chemical Identifiers

CAS 19621-92-2
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00192220
InChI Key VRCWSYYXUCKEED-UHFFFAOYSA-N
Synonym 6-hydroxypicolinic acid, 6-hydroxy-2-pyridinecarboxylic acid, 6-hydroxypyridine-2-carboxylic acid, 6-oxo-1,6-dihydropyridine-2-carboxylic acid, 6-carboxy-2-hydroxy-pyridine, 6-hydroxy-2-pyridine carboxylic acid, 6-hydroxy-2-pyridinecarboxylicacid, 2-pyridinecarboxylic acid, 1,6-dihydro-6-oxo, 2-pyridinecarboxylicacid, 1,6-dihydro-6-oxo, 2-pyridinecarboxylic acid,1,6-dihydro-6-oxo
PubChem CID 242721
IUPAC Name 6-oxo-1H-pyridine-2-carboxylic acid
SMILES C1=CC(=O)NC(=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H087625G View Documents TCI America
H087625G
25g Each for $305.56
 
H08765G View Documents TCI America
H08765G
5g Each for $100.83
 
Specifications

Chemical Identifiers

19621-92-2
139.11
VRCWSYYXUCKEED-UHFFFAOYSA-N
242721
C1=CC(=O)NC(=C1)C(=O)O
C6H5NO3
MFCD00192220
6-hydroxypicolinic acid, 6-hydroxy-2-pyridinecarboxylic acid, 6-hydroxypyridine-2-carboxylic acid, 6-oxo-1,6-dihydropyridine-2-carboxylic acid, 6-carboxy-2-hydroxy-pyridine, 6-hydroxy-2-pyridine carboxylic acid, 6-hydroxy-2-pyridinecarboxylicacid, 2-pyridinecarboxylic acid, 1,6-dihydro-6-oxo, 2-pyridinecarboxylicacid, 1,6-dihydro-6-oxo, 2-pyridinecarboxylic acid,1,6-dihydro-6-oxo
6-oxo-1H-pyridine-2-carboxylic acid

Specifications

Undesignated
19621-92-2
MFCD00192220
VRCWSYYXUCKEED-UHFFFAOYSA-N
6-oxo-1H-pyridine-2-carboxylic acid
242721
Crystal-Powder at 20°C
6-Hydroxy-2-pyridinecarboxylic Acid
25g
C6H5NO3
6-hydroxypicolinic acid, 6-hydroxy-2-pyridinecarboxylic acid, 6-hydroxypyridine-2-carboxylic acid, 6-oxo-1,6-dihydropyridine-2-carboxylic acid, 6-carboxy-2-hydroxy-pyridine, 6-hydroxy-2-pyridine carboxylic acid, 6-hydroxy-2-pyridinecarboxylicacid, 2-pyridinecarboxylic acid, 1,6-dihydro-6-oxo, 2-pyridinecarboxylicacid, 1,6-dihydro-6-oxo, 2-pyridinecarboxylic acid,1,6-dihydro-6-oxo
C1=CC(=O)NC(=C1)C(=O)O
139.11
139.11
≥97.0% (T)
Safety and Handling

Safety and Handling

TSCA : No