missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Hydroxy-2-pyridinecarboxylic Acid 97.0+%, TCI America™
Supplier: TCI America H087625G
Specifications
6-Hydroxy-2-pyridinecarboxylic Acid | |
C6H5NO3 | |
25 g | |
VRCWSYYXUCKEED-UHFFFAOYSA-N | |
6-oxo-1H-pyridine-2-carboxylic acid | |
242721 | |
≥97.0% (T) |
19621-92-2 | |
MFCD00192220 | |
6-hydroxypicolinic acid, 6-hydroxy-2-pyridinecarboxylic acid, 6-hydroxypyridine-2-carboxylic acid, 6-oxo-1,6-dihydropyridine-2-carboxylic acid, 6-carboxy-2-hydroxy-pyridine, 6-hydroxy-2-pyridine carboxylic acid, 6-hydroxy-2-pyridinecarboxylicacid, 2-pyridinecarboxylic acid, 1,6-dihydro-6-oxo, 2-pyridinecarboxylicacid, 1,6-dihydro-6-oxo, 2-pyridinecarboxylic acid,1,6-dihydro-6-oxo | |
C1=CC(=O)NC(=C1)C(=O)O | |
139.11 | |
139.11 | |
Crystalline Powder |
Chemical Identifiers
19621-92-2 | |
139.11 | |
VRCWSYYXUCKEED-UHFFFAOYSA-N | |
242721 | |
C1=CC(=O)NC(=C1)C(=O)O |
C6H5NO3 | |
MFCD00192220 | |
6-hydroxypicolinic acid, 6-hydroxy-2-pyridinecarboxylic acid, 6-hydroxypyridine-2-carboxylic acid, 6-oxo-1,6-dihydropyridine-2-carboxylic acid, 6-carboxy-2-hydroxy-pyridine, 6-hydroxy-2-pyridine carboxylic acid, 6-hydroxy-2-pyridinecarboxylicacid, 2-pyridinecarboxylic acid, 1,6-dihydro-6-oxo, 2-pyridinecarboxylicacid, 1,6-dihydro-6-oxo, 2-pyridinecarboxylic acid,1,6-dihydro-6-oxo | |
6-oxo-1H-pyridine-2-carboxylic acid |
Safety and Handling
TSCA : No