Please login to your online account to display your discounted pricing

6-Hydroxy-2-naphthoic acid, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  347650250

 View more versions of this product

Catalog No. AC347650250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 188.18
Quantity 25g
Color Pale brown
EINECS Number 240-759-7
TSCA TSCA
RTECS Number QN3722500
Melting Point 240° to 250°C
CAS Max % 100.0
Chemical Name or Material 6-Hydroxy-2-naphthoic acid
Packaging Glass bottle
CAS 16712-64-4
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C11H8O3
Linear Formula HOC10H6COOH
MDL Number MFCD00060070

Specifications

Chemical Name or Material 6-Hydroxy-2-naphthoic acid
CAS 16712-64-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H8O3
Linear Formula HOC10H6COOH
Formula Weight 188.18
MDL Number MFCD00060070
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents