Please login to your online account to display your discounted pricing

6-Hydroxy-2-naphthoic acid, 98%, ACROS Organics™

$311.33 - $311.33

Chemical Identifiers

Chemical Name or Material 6-Hydroxy-2-naphthoic acid
CAS 16712-64-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H8O3
Linear Formula HOC10H6COOH
Formula Weight 188.18
MDL Number MFCD00060070
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC347650050

  SDS
fisher scientific acros
347650050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC347650250

  SDS
thermo fisher scientific
347650250
Glass bottle 25g Each for $311.33
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 188.18
Quantity 5g
Color Pale brown
EINECS Number 240-759-7
TSCA TSCA
RTECS Number QN3722500
Melting Point 240° to 250°C
CAS Max % 100.0
Chemical Name or Material 6-Hydroxy-2-naphthoic acid
Packaging Glass bottle
Molecular Formula C11H8O3
Linear Formula HOC10H6COOH
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD00060070
CAS 16712-64-4
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.