missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Hydroxy-2-naphthaldehyde, TCI America™

$73.33 - $247.50

Chemical Identifiers

CAS 78119-82-1
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
MDL Number MFCD03427537
InChI Key PRYNJOJHKYNLIS-UHFFFAOYSA-N
Synonym 2-Naphthol-6-carboxaldehyde
PubChem CID 2764110
IUPAC Name 6-hydroxynaphthalene-2-carbaldehyde
SMILES C1=CC2=C(C=CC(=C2)O)C=C1C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H12455G View Documents TCI America
H12455G
5g Each for $73.33
 
H124525G View Documents TCI America
H124525G
25g Each for $247.50
 
Specifications

Chemical Identifiers

78119-82-1
172.183
PRYNJOJHKYNLIS-UHFFFAOYSA-N
2764110
C1=CC2=C(C=CC(=C2)O)C=C1C=O
C11H8O2
MFCD03427537
2-Naphthol-6-carboxaldehyde
6-hydroxynaphthalene-2-carbaldehyde

Specifications

Undesignated
5g
C11H8O2
2-Naphthol-6-carboxaldehyde
C1=CC2=C(C=CC(=C2)O)C=C1C=O
172.183
172.18
≥98.0% (GC,T)
183°C
78119-82-1
MFCD03427537
PRYNJOJHKYNLIS-UHFFFAOYSA-N
6-hydroxynaphthalene-2-carbaldehyde
2764110
Crystal-Powder at 20°C
6-Hydroxy-2-naphthaldehyde
Safety and Handling

Safety and Handling

TSCA : No