missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Hydroxy-2-naphthaldehyde 98.0+%, TCI America™

Manufacturer:  Tci America H12455G

 View more versions of this product

Catalog No. H12455G



Specifications

Specifications

6-Hydroxy-2-naphthaldehyde
183°C
78119-82-1
MFCD03427537
PRYNJOJHKYNLIS-UHFFFAOYSA-N
6-hydroxynaphthalene-2-carbaldehyde
2764110
Crystal-Powder at 20°C
Undesignated
5g
C11H8O2
2-Naphthol-6-carboxaldehyde
C1=CC2=C(C=CC(=C2)O)C=C1C=O
172.183
172.18
≥98.0% (GC,T)

Chemical Identifiers

78119-82-1
172.183
PRYNJOJHKYNLIS-UHFFFAOYSA-N
2764110
C1=CC2=C(C=CC(=C2)O)C=C1C=O
C11H8O2
MFCD03427537
2-Naphthol-6-carboxaldehyde
6-hydroxynaphthalene-2-carbaldehyde
Safety and Handling

Safety and Handling

TSCA : No

Documents
Promotions

Promotions