Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Hydroxy-2-methylthio-4(3H)-pyrimidinone, 97%

$80.00 - $248.62

Chemical Identifiers

Chemical Name or Material 6-Hydroxy-2-methylthio-4(3H)-pyrimidinone
CAS 1979-98-2
Molecular Formula C5H6N2O2S
Formula Weight 158.18
MDL Number MFCD00006087
Synonym 4,6-Dihydroxy-2-(methylthio)pyrimidine; 2-Methylthio-4,6-pyrimidinedione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2870814

  SDS
alfa aesar™
H2870814
25g Each for $248.62

AAH2870806

  SDS
alfa aesar™
H2870806
5g Each for $80.00
Description & Specifications

Specifications

Formula Weight 158.18
Synonym 4,6-Dihydroxy-2-(methylthio)pyrimidine; 2-Methylthio-4,6-pyrimidinedione
Melting Point >300°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Chemical Name or Material 6-Hydroxy-2-methylthio-4(3H)-pyrimidinone
CAS 1979-98-2
Quantity 25g
Assay 97%
Molecular Formula C5H6N2O2S
MDL Number MFCD00006087
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.