missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Hydroxy-2-methylquinoline 98.0+%, TCI America™
$170.87 - $499.37
Chemical Identifiers
CAS | 613-21-8 |
---|---|
Molecular Formula | C10H9NO |
Molecular Weight (g/mol) | 159.188 |
MDL Number | MFCD00047600 |
InChI Key | USSQQASIZNTRAJ-UHFFFAOYSA-N |
Synonym | 6-hydroxy-2-methylquinoline, 6-hydroxyquinaldine, 2-methyl-6-quinolinol, 6-quinolinol, 2-methyl, 2-methyl-6-hydroxyquinoline, 2-methyl-quinolin-6-ol, acmc-1bit8, 2-methyl-6-quinolinol #, 6-hydroxy-2-methyl quionoline, 6-hydroxy-2-methylquinoline 1g |
PubChem CID | 594395 |
IUPAC Name | 2-methylquinolin-6-ol |
SMILES | CC1=NC2=C(C=C1)C=C(C=C2)O |
Chemical Identifiers
613-21-8 | |
159.188 | |
USSQQASIZNTRAJ-UHFFFAOYSA-N | |
594395 | |
CC1=NC2=C(C=C1)C=C(C=C2)O |
C10H9NO | |
MFCD00047600 | |
6-hydroxy-2-methylquinoline, 6-hydroxyquinaldine, 2-methyl-6-quinolinol, 6-quinolinol, 2-methyl, 2-methyl-6-hydroxyquinoline, 2-methyl-quinolin-6-ol, acmc-1bit8, 2-methyl-6-quinolinol #, 6-hydroxy-2-methyl quionoline, 6-hydroxy-2-methylquinoline 1g | |
2-methylquinolin-6-ol |
Specifications
613-21-8 | |
C10H9NO | |
1 g | |
USSQQASIZNTRAJ-UHFFFAOYSA-N | |
2-methylquinolin-6-ol | |
594395 | |
≥98.0% (GC,T) | |
6-Hydroxy-2-methylquinoline |
216°C | |
MFCD00047600 | |
6-hydroxy-2-methylquinoline, 6-hydroxyquinaldine, 2-methyl-6-quinolinol, 6-quinolinol, 2-methyl, 2-methyl-6-hydroxyquinoline, 2-methyl-quinolin-6-ol, acmc-1bit8, 2-methyl-6-quinolinol #, 6-hydroxy-2-methyl quionoline, 6-hydroxy-2-methylquinoline 1g | |
CC1=NC2=C(C=C1)C=C(C=C2)O | |
159.188 | |
159.19 | |
Crystalline Powder |
Safety and Handling
TSCA : No