Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Hydroxy-2,8-dimercaptopurine, 98%

$31.09 - $466.41

Chemical Identifiers

Chemical Name or Material 6-Hydroxy-2,8-dimercaptopurine
CAS 15986-32-0
Molecular Formula C5H4N4OS2
Formula Weight 200.24
MDL Number MFCD00047144
Synonym 2,8-Dimercapto-6-hydroxypurine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1717803

 
alfa aesar™
A1717803
1g Each for $31.09

AAA1717806

 
alfa aesar™
A1717806
5g Each for $116.07

AAA1717814

 
alfa aesar™
A1717814
25g Each for $466.41
Description & Specifications

Specifications

Formula Weight 200.24
Chemical Name or Material 6-Hydroxy-2,8-dimercaptopurine
Melting Point >320°C
Quantity 1g
Assay 98%
Molecular Formula C5H4N4OS2
MDL Number MFCD00047144
CAS 15986-32-0
Synonym 2,8-Dimercapto-6-hydroxypurine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UO9050000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.