Please login to your online account to display your discounted pricing

6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 6-Hydroxy-2, 3-dihydrobenzo[b]furan-3-one
CAS 6272-26-0
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H6O3
Formula Weight 150.13
MDL Number MFCD00068174
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW02738DA

 
fisher scientific company
SEW02738DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 150.13
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 6-Hydroxy-2, 3-dihydrobenzo[b]furan-3-one
Assay Percent Range 97%
Molecular Formula C8H6O3
CAS 6272-26-0
Packaging Amber Glass Bottle
MDL Number MFCD00068174
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.