Learn More
6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L10951MD
Description
6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one, is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one, is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one | |
240°C to 242°C | |
MFCD00068174 | |
121444 | |
Slightly soluble in water. | |
OC1=CC=C2C(=O)COC2=C1 | |
150.13 | |
150.14 |
6272-26-0 | |
C8H6O3 | |
250 mg | |
6-hydroxybenzofuran-3 2h-one, 6-hydroxy-2h-benzofuran-3-one, 6-hydroxy-2,3-dihydrobenzo b furan-3-one, 6-hydroxy-1-benzofuran-3 2h-one, 6-hydroxybenzofuran-3-one, 3 2h-benzofuranone, 6-hydroxy, 6-hydroxy-3-coumaranone, 6-hydroxy-2,3-dihydro-1-benzofuran-3-one, 6-hydroxy-3 2h-benzofuranone, 6-hydroxy-2h-1-benzofuran-3-one | |
GBDMODVZBPFQKI-UHFFFAOYSA-N | |
6-hydroxy-2,3-dihydro-1-benzofuran-3-one | |
235717 | |
97% |
Chemical Identifiers
6272-26-0 | |
150.13 | |
GBDMODVZBPFQKI-UHFFFAOYSA-N | |
235717 | |
OC1=CC=C2C(=O)COC2=C1 |
C8H6O3 | |
MFCD00068174 | |
6-hydroxybenzofuran-3 2h-one, 6-hydroxy-2h-benzofuran-3-one, 6-hydroxy-2,3-dihydrobenzo b furan-3-one, 6-hydroxy-1-benzofuran-3 2h-one, 6-hydroxybenzofuran-3-one, 3 2h-benzofuranone, 6-hydroxy, 6-hydroxy-3-coumaranone, 6-hydroxy-2,3-dihydro-1-benzofuran-3-one, 6-hydroxy-3 2h-benzofuranone, 6-hydroxy-2h-1-benzofuran-3-one | |
6-hydroxy-2,3-dihydro-1-benzofuran-3-one |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only