missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
6-Hydroxy-1-indanone, TCI America™
Chemical Identifiers
CAS |
62803-47-8 |
Molecular Formula |
C9H8O2 |
Molecular Weight (g/mol) |
148.161 |
MDL Number |
MFCD00513530 |
InChI Key |
MOANRQDXNNXOLW-UHFFFAOYSA-N |
PubChem CID |
7020659 |
IUPAC Name |
6-hydroxy-2,3-dihydroinden-1-one |
SMILES |
C1CC(=O)C2=C1C=CC(=C2)O |
View More Specs
Products
2
Specifications
Chemical Identifiers
|
62803-47-8 |
|
148.161 |
|
MOANRQDXNNXOLW-UHFFFAOYSA-N |
|
6-hydroxy-2,3-dihydroinden-1-one |
|
C9H8O2 |
|
MFCD00513530 |
|
7020659 |
|
C1CC(=O)C2=C1C=CC(=C2)O |
Specifications
|
Yellow |
|
1g |
|
C9H8O2 |
|
MOANRQDXNNXOLW-UHFFFAOYSA-N |
|
6-hydroxy-2,3-dihydroinden-1-one |
|
7020659 |
|
Crystal-Powder at 20°C |
|
6-Hydroxy-1-indanone |
|
157°C |
|
62803-47-8 |
|
MFCD00513530 |
|
C1CC(=O)C2=C1C=CC(=C2)O |
|
148.161 |
|
148.16 |
|
≥98.0% (GC) |
Safety and Handling