Please login to your online account to display your discounted pricing

6-Hydrazino-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 6-Hydrazino-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine
CAS 175202-00-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H13N5
Formula Weight 191.24
MDL Number MFCD00067969
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM02421CB

 
fisher scientific company
KM02421CB
Amber glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KM02421DA

 
fisher scientific company
KM02421DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.24
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material 6-Hydrazino-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine
Molecular Formula C9H13N5
Packaging Amber glass bottle
MDL Number MFCD00067969
CAS 175202-00-3
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%