Please login to your online account to display your discounted pricing

6-Hydrazino-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine, 97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  KM02421DA

 View more versions of this product

Catalog No. KM02421DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 191.24
Quantity 1g
Assay 97%
CAS Min % 97.0
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Molecular Formula C9H13N5
MDL Number MFCD00067969
CAS 175202-00-3
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Chemical Name or Material 6-Hydrazino-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine
Packaging Amber glass bottle

Specifications

Chemical Name or Material 6-Hydrazino-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine
CAS 175202-00-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H13N5
Formula Weight 191.24
MDL Number MFCD00067969
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.