missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ 6-Histidine (Epitope Tag) Ab-1, Mouse Monoclonal Antibody

Recommended for Immunofluorescence and Affinity Purification, Thermo Scientific™ 6-Histidine (Epitope Tag) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

$243.29 - $1,008.08

Specifications

Antigen 6-Histidine (Epitope Tag) Ab-1
Clone 4D11
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS667P Thermo Scientific™ Lab Vision™
MS667P
200μg/mL 1mL Each for $959.45
MS667P0 Thermo Scientific™ Lab Vision™
MS667P0
200μg/mL 100μL Each for $243.29
MS667P1 Thermo Scientific™ Lab Vision™
MS667P1
200μg/mL 500μL Each for $679.02
MS667P1ABX Thermo Scientific™ Lab Vision™
MS667P1ABX
1mg/mL 100μL Each for $765.80
MS667PABX Thermo Scientific™ Lab Vision™
MS667PABX
1mg/mL 200μL Each for $1,008.08
Specifications

Description

Recommended for Immunofluorescence and Affinity Purification, Thermo Scientific™ 6-Histidine (Epitope Tag) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Insertion of a 6-Histidine tag creates a stable fusion product that does not interfere with the bioactivity of the protein or with the biodistribution of the His-tagged product. Such proteins are purified by immobilized metal ion affinity chromatography which makes use of its high affinity for transition metal ion.

Host Species: Mouse

Clone: 4D11

Isotype: IgG2a/κ

Species Reactivity: All species.

Epitope: HHHHHHGS

Immunogen: A His-tagged recombinant protein

Positive Control: Hexa-histidine fusion protein

Cellular Localization: Depends upon the localization of the parent protein tagged with hexa-histidine

Recommended for:

  • Western Blotting (Not verified)
  • Immunofluorescence
  • Affinity Purification

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

6-Histidine (Epitope Tag) Ab-1
Immunofluorescence
Unlabeled
Murine
Epitope Tags
A His-tagged recombinant protein
4D11
Monoclonal
Concentrated
RUO
Primary
IgG2a kappa