missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ 6-Histidine (Epitope Tag) Ab-1, Mouse Monoclonal Antibody

Recommended for Immunofluorescence and Affinity Purification, Thermo Scientific™ 6-Histidine (Epitope Tag) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS667P1ABX

 View more versions of this product

Catalog No. MS667P1ABX


 

Specifications

Description

Recommended for Immunofluorescence and Affinity Purification, Thermo Scientific™ 6-Histidine (Epitope Tag) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Insertion of a 6-Histidine tag creates a stable fusion product that does not interfere with the bioactivity of the protein or with the biodistribution of the His-tagged product. Such proteins are purified by immobilized metal ion affinity chromatography which makes use of its high affinity for transition metal ion.

Host Species: Mouse

Clone: 4D11

Isotype: IgG2a/κ

Species Reactivity: All species.

Epitope: HHHHHHGS

Immunogen: A His-tagged recombinant protein

Positive Control: Hexa-histidine fusion protein

Cellular Localization: Depends upon the localization of the parent protein tagged with hexa-histidine

Recommended for:

  • Western Blotting (Not verified)
  • Immunofluorescence
  • Affinity Purification

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

6-Histidine (Epitope Tag) Ab-1
Monoclonal
1mg/mL
Concentrated
A His-tagged recombinant protein
100μL
Epitope Tags
Monoclonal
Immunofluorescence
4D11
Unlabeled
Murine
IgG2a kappa
RUO
Primary
All