missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Hexanoyl-D-erythro-sphingosine, 99+%, synthetical, ACROS Organics™

$570.94 - $570.94

Chemical Identifiers

CAS 124753-97-5
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Molecular Formula C24H47NO3
Formula Weight 397.64
MDL Number MFCD00171385
Synonym Ceramide C6
Chemical Name or Material 6-Hexanoyl-D-erythro-sphingosine, 99+%
Assay ≥99.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC346220500 SDS Thermo Fisher Scientific
346220500
Glass bottle 50mg Each for $570.94
Specifications Description & Specifications

Chemical Identifiers

124753-97-5
100.0
C24H47NO3
MFCD00171385
6-Hexanoyl-D-erythro-sphingosine, 99+%
99.0
99+%
397.64
Ceramide C6
≥99.0%

Specifications

Powder
50mg
100.0
Authentic
Glass bottle
124753-97-5
MFCD00171385
≥99.0%
397.64
White
99.0
Ceramide C6
99+%
C24H47NO3
6-Hexanoyl-D-erythro-sphingosine, 99+%