Please login to your online account to display your discounted pricing

6-Hexanoyl-D-erythro-sphingosine, 99+%, synthetical, ACROS Organics™


Quantity 50mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  346220500

 View more versions of this product

Catalog No. AC346220500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Powder
Formula Weight 397.64
Quantity 50mg
Color White
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material 6-Hexanoyl-D-erythro-sphingosine
Infrared Spectrum Authentic
Name Note 99+%
Packaging Glass bottle
Assay Percent Range 99+%
Molecular Formula C24H47NO3
MDL Number MFCD00171385
CAS 124753-97-5
Synonym Ceramide C6

Specifications

Chemical Name or Material 6-Hexanoyl-D-erythro-sphingosine
Name Note 99+%
CAS 124753-97-5
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C24H47NO3
Formula Weight 397.64
MDL Number MFCD00171385
Synonym Ceramide C6