Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Hexadecanone, 97%

$179.08 - $179.08

Chemical Identifiers

Chemical Name or Material 6-Hexadecanone
CAS 57661-23-1
Molecular Formula C16H32O
Formula Weight 240.43g/mol
MDL Number MFCD00051549
Synonym n-Amyl n-decyl ketone; n-Decyl n-pentyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2515406

 
alfa aesar™
B25154-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2515414

 
alfa aesar™
B25154-14
25g Each for $179.08
Description & Specifications

Specifications

Melting Point 34° to 38°C
Quantity 5g
Chemical Name or Material 6-Hexadecanone
CAS 57661-23-1
Assay 97%
Molecular Formula C16H32O
Formula Weight 240.43g/mol
MDL Number MFCD00051549
Synonym n-Amyl n-decyl ketone; n-Decyl n-pentyl ketone