Learn More
6-Heptynoic acid, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5351906
Description
6-Heptynoic acid is used as a precursor for the the preparation of boradiazaindacenes (BODIPY dyes) by reacting with oxalylchloirde. It is used to prepare 4,4-difluoro-8-(hept-6-yne)-3,5-di(2-(4-methoxyphenyl))-4-bora-3a,4a-diaza-sindacene by reacting with 2-(4-methoxyphenyl)-1H-pyrrole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Heptynoic acid is used as a precursor for the the preparation of boradiazaindacenes (BODIPY dyes) by reacting with oxalylchloirde. It is used to prepare 4,4-difluoro-8-(hept-6-yne)-3,5-di(2-(4-methoxyphenyl))-4-bora-3a,4a-diaza-sindacene by reacting with 2-(4-methoxyphenyl)-1H-pyrrole.
Solubility
Miscible with dimethylformamide.
Notes
Incompatible with oxidizing agents and bases
Specifications
6-Heptynoic acid | |
0.997 | |
113°C (235°F) | |
1.451 | |
MFCD00239430 | |
UN3265 | |
6-heptynoic acid, 6-heptynoicacid, 6-heptynoic alcohol, ksc222a4l, ofcpmjgtzuvusm-uhfffaoysa | |
OFCPMJGTZUVUSM-UHFFFAOYSA-N | |
hept-6-ynoic acid | |
4377950 | |
95% |
30964-00-2 | |
93°C to 94°C (1 mmHg) | |
C7H10O2 | |
HC≡C(CH2)4CO2H | |
5 g | |
1747024 | |
Miscible with dimethylformamide. | |
OC(=O)CCCCC#C | |
126.16 | |
126.16 |
Chemical Identifiers
30964-00-2 | |
126.16 | |
OFCPMJGTZUVUSM-UHFFFAOYSA-N | |
4377950 | |
OC(=O)CCCCC#C |
C7H10O2 | |
MFCD00239430 | |
6-heptynoic acid, 6-heptynoicacid, 6-heptynoic alcohol, ksc222a4l, ofcpmjgtzuvusm-uhfffaoysa | |
hept-6-ynoic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only