missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Heptynenitrile 98.0+%, TCI America™
$341.76 - $1134.76
Chemical Identifiers
CAS | 15295-69-9 |
---|---|
Molecular Formula | C7H9N |
Molecular Weight (g/mol) | 107.156 |
InChI Key | BIGRRVLEKCQFHS-UHFFFAOYSA-N |
Synonym | 6-Cyano-1-hexyne |
PubChem CID | 11768526 |
IUPAC Name | hept-6-ynenitrile |
SMILES | C#CCCCCC#N |
Chemical Identifiers
15295-69-9 | |
107.156 | |
6-Cyano-1-hexyne | |
hept-6-ynenitrile |
C7H9N | |
BIGRRVLEKCQFHS-UHFFFAOYSA-N | |
11768526 | |
C#CCCCCC#N |
Specifications
15295-69-9 | |
93°C | |
5 g | |
6-Cyano-1-hexyne | |
C#CCCCCC#N | |
107.156 | |
107.16 | |
Liquid |
Yellow | |
C7H9N | |
3276 | |
BIGRRVLEKCQFHS-UHFFFAOYSA-N | |
hept-6-ynenitrile | |
11768526 | |
≥98.0% (GC) | |
6-Heptynenitrile |
Safety and Handling
TSCA : No