missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Heptyn-1-ol, 95%

$268.32 - $1,074.85

Chemical Identifiers

CAS 63478-76-2
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00049198
InChI Key BVRCLEXKQNWTDK-UHFFFAOYSA-N
Synonym 6-heptyn-1-ol, 7-hydroxyhept-1-yne, pubchem13080, ho ch2 5c=ch, ksc352m8p
PubChem CID 11007849
IUPAC Name hept-6-yn-1-ol
SMILES C#CCCCCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6150903 View Documents Alfa Aesar™
H6150903
1g Each for $268.32
 
AAH6150906 View Documents Alfa Aesar™
H6150906
5g Each for $1,074.85
 
Description

Description

Applications
6-Heptyn-1-ol is used as a reagent in in the preparation of many cyclic compounds. It is also used as a Intermediate in organic synthesis.

Solubility
Soluble in chloroform, dichloromethane and methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Chemical Identifiers

63478-76-2
112.172
BVRCLEXKQNWTDK-UHFFFAOYSA-N
11007849
C#CCCCCCO
C7H12O
MFCD00049198
6-heptyn-1-ol, 7-hydroxyhept-1-yne, pubchem13080, ho ch2 5c=ch, ksc352m8p
hept-6-yn-1-ol

Specifications

1g
C7H12O
MFCD00049198
BVRCLEXKQNWTDK-UHFFFAOYSA-N
hept-6-yn-1-ol
11007849
95%
63478-76-2
HC≡C(CH2)5OH
6-heptyn-1-ol, 7-hydroxyhept-1-yne, pubchem13080, ho ch2 5c=ch, ksc352m8p
C#CCCCCCO
112.172
112.17
6-Heptyn-1-ol
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C