missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Heptenoic acid, 98%

Manufacturer:  Alfa Aesar™ L1314406

 View more versions of this product

Catalog No. AAL1314406



Description

Description

Applications
6-Heptenoic acid is used in the preparation of 6-(iodomethyl)-hexanolide through iodo-lactonization.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
Specifications

Specifications

6-Heptenoic acid
Unpleasant
~-6°C
1119-60-4
MFCD00014382
1747265
RWNJOXUVHRXHSD-UHFFFAOYSA-N
hept-6-enoic acid
70705
128.17
>110°C (230°F)
222°C to 224°C
5g
C7H12O2
UN3265
6-heptenoic acid, epsilon-heptenoic acid, rarechem al bo 1361, 6-heptenoic acids, hept-6-enoic acids, acmc-20alyp, epsilon-heptenoic acids, 6-heptenoic acid, 6-heptenoic acid 1g
C=CCCCCC(=O)O
128.171
CHEBI:38363
98%

Chemical Identifiers

1119-60-4
128.171
RWNJOXUVHRXHSD-UHFFFAOYSA-N
70705
hept-6-enoic acid
C7H12O2
MFCD00014382
6-heptenoic acid, epsilon-heptenoic acid, rarechem al bo 1361, 6-heptenoic acids, hept-6-enoic acids, acmc-20alyp, epsilon-heptenoic acids, 6-heptenoic acid, 6-heptenoic acid 1g
CHEBI:38363
C=CCCCCC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Documents