Please login to your online account to display your discounted pricing

6-Fluorotryptamine hydrochloride, 99%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  229245000

 View more versions of this product

Catalog No. AC229245000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 214.67
Physical Form Fine Crystalline Powder
Quantity 500mg
Color White to slightly beige
Melting Point 233° to 236°C
EINECS Number 259-533-4
CAS Max % 100.0
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Chemical Name or Material 6-Fluorotryptamine hydrochloride
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99%
Molecular Formula C10 H11 F N2 . H Cl
MDL Number MFCD00012687
Name Note 99%
CAS 55206-24-1

Specifications

Chemical Name or Material 6-Fluorotryptamine hydrochloride
Name Note 99%
CAS 55206-24-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10 H11 F N2 . H Cl
Formula Weight 214.67
MDL Number MFCD00012687
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents