Please login to your online account to display your discounted pricing

6-Fluorotryptamine hydrochloride, 99%, ACROS Organics™


Quantity 100mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  229241000

 View more versions of this product

Catalog No. AC229241000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Fine Crystalline Powder
Formula Weight 214.67
Quantity 100mg
Color White to slightly beige
EINECS Number 259-533-4
Melting Point 233° to 236°C
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
CAS 55206-24-1
Packaging Glass bottle
Assay Percent Range 99%
MDL Number MFCD00012687
Name Note 99%
Chemical Name or Material 6-Fluorotryptamine hydrochloride
Molecular Formula C10 H11 F N2 . H Cl

Specifications

Chemical Name or Material 6-Fluorotryptamine hydrochloride
Name Note 99%
CAS 55206-24-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10 H11 F N2 . H Cl
Formula Weight 214.67
MDL Number MFCD00012687
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents