Please login to your online account to display your discounted pricing

6-Fluorotryptamine hydrochloride, 99%, ACROS Organics™

$594.62 - $594.62

Chemical Identifiers

Chemical Name or Material 6-Fluorotryptamine hydrochloride
Name Note 99%
CAS 55206-24-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10 H11 F N2 . H Cl
Formula Weight 214.67
MDL Number MFCD00012687
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC229245000

  SDS
thermo fisher scientific
229245000
Glass bottle 500mg Each for $594.62

AC229241000

  SDS
thermo fisher scientific
229241000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Fine Crystalline Powder
Formula Weight 214.67
Quantity 500mg
Color White to slightly beige
EINECS Number 259-533-4
Melting Point 233° to 236°C
CAS Max % 100.0
CAS Min % 98.5
Chemical Name or Material 6-Fluorotryptamine hydrochloride
Name Note 99%
Assay Percent Range 99%
Molecular Formula C10 H11 F N2 . H Cl
MDL Number MFCD00012687
CAS 55206-24-1
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.