Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Fluorosalicylic acid, 96%

$55.45 - $147.66

Chemical Identifiers

Chemical Name or Material 6-Fluorosalicylic acid
CAS 67531-86-6
Molecular Formula C7H5FO3
Formula Weight 156.11
MDL Number MFCD00052179
Beilstein 7111497
Synonym 6-Fluoro-2-hydroxybenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1090703

  SDS
alfa aesar™
L1090703
1g Each for $147.66

AAL10907MD

  SDS
alfa aesar™
L10907MD
250mg Each for $55.45
Description & Specifications

Specifications

Formula Weight 156.11
Molecular Formula C7H5FO3
MDL Number MFCD00052179
Assay 96%
Chemical Name or Material 6-Fluorosalicylic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 6-Fluoro-2-hydroxybenzoic acid
CAS 67531-86-6
Health Hazard 1 Warning
Melting Point 159° to 161°C
Quantity 1g
Beilstein 7111497
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.