missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Fluorosalicylic acid, 96%

Manufacturer:  Alfa Aesar™ L1090703

 View more versions of this product

Catalog No. AAL1090703



Specifications

Specifications

6-Fluorosalicylic acid
1g
C7H5FO3
7111497
BCEKGWWLVKXZKK-UHFFFAOYSA-N
2-fluoro-6-hydroxybenzoic acid
586777
96%
159°C to 161°C
67531-86-6
MFCD00052179
6-fluorosalicylic acid, 6-fluoro-2-hydroxybenzoic acid, 2-fluoro-6-hydroxybenzoicacid, 6-fluorosalicylicacid, benzoic acid, 2-fluoro-6-hydroxy, 2-fluoro-6-hydoxybenzoic acid, pubchem4958, acmc-1baav, 2-carboxy-3-fluorophenol
C1=CC(=C(C(=C1)F)C(=O)O)O
156.112
156.11

Chemical Identifiers

67531-86-6
156.112
BCEKGWWLVKXZKK-UHFFFAOYSA-N
586777
C1=CC(=C(C(=C1)F)C(=O)O)O
C7H5FO3
MFCD00052179
6-fluorosalicylic acid, 6-fluoro-2-hydroxybenzoic acid, 2-fluoro-6-hydroxybenzoicacid, 6-fluorosalicylicacid, benzoic acid, 2-fluoro-6-hydroxy, 2-fluoro-6-hydoxybenzoic acid, pubchem4958, acmc-1baav, 2-carboxy-3-fluorophenol
2-fluoro-6-hydroxybenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents