Please login to your online account to display your discounted pricing

6-Fluoropyridine-3-carboxylic acid, 95%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC436830050

 View more versions of this product

Catalog No. AC436830050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 141.1
Quantity 5g
Color White to off-white
CAS Min % 94.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Range 95%
Molecular Formula C6H4FNO2
Chemical Name or Material 6-Fluoropyridine-3-carboxylic acid
CAS 403-45-2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Synonym 6-Fluoronicotinic acid
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 6-Fluoropyridine-3-carboxylic acid
CAS 403-45-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 95%
Molecular Formula C6H4FNO2
Formula Weight 141.1
Synonym 6-Fluoronicotinic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents