Please login to your online account to display your discounted pricing

6-Fluoropyridine-3-carboxylic acid, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 6-Fluoropyridine-3-carboxylic acid
CAS 403-45-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 95%
Molecular Formula C6H4FNO2
Formula Weight 141.1
Synonym 6-Fluoronicotinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC436830010

  SDS
thermo fisher scientific
AC436830010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC436830050

  SDS
thermo fisher scientific
AC436830050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 141.1
Quantity 1g
Color White to off-white
CAS Min % 94.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 6-Fluoropyridine-3-carboxylic acid
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C6H4FNO2
CAS 403-45-2
Synonym 6-Fluoronicotinic acid
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.