missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Fluoroindole, 98%

$110.55 - $1,392.70

Chemical Identifiers

CAS 399-51-9
Molecular Formula C8H6FN
Formula Weight 135.14
MDL Number MFCD00056933
Beilstein 112192
Chemical Name or Material 6-Fluoroindole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2463314 SDS Alfa Aesar™
B2463314
25g Each for $1,392.70
AAB2463303 SDS Alfa Aesar™
B2463303
1g Each for $110.55
AAB2463306 SDS Alfa Aesar™
B2463306
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
6-Fluoroindole acts as a reagent in the synthesis of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles, which acts as a potential anticancer immunomodulator. It is also employed as an antifungal and antibacterial agent. Further, it serves as a potent selective serotonin reuptake inhibitor. In addition to this, it is used as a inhibitor of HIV-1 attachment.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in a cool place.Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.
Description & Specifications

Chemical Identifiers

399-51-9
135.14
112192
C8H6FN
MFCD00056933
6-Fluoroindole

Specifications

74° to 77°C
399-51-9
135.14
112192
98%
25g
C8H6FN
MFCD00056933
6-Fluoroindole

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning