Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Fluorochroman-4-one, 97%

$54.69 - $173.28

Chemical Identifiers

Chemical Name or Material 6-Fluorochroman-4-one
CAS 66892-34-0
Molecular Formula C9H7FO2
Formula Weight 166.15
MDL Number MFCD00038654
Beilstein 4245390
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1120906

  SDS
alfa aesar™
L1120906
5g Each for $173.28

AAL1120903

  SDS
alfa aesar™
L1120903
1g Each for $54.69
Description & Specifications

Specifications

Formula Weight 166.15
Chemical Name or Material 6-Fluorochroman-4-one
Assay 97%
Molecular Formula C9H7FO2
MDL Number MFCD00038654
CAS 66892-34-0
Beilstein 4245390
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 114° to 115°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.