missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Fluorochroman-4-one, 97%

Manufacturer:  Alfa Aesar™ L1120906

 View more versions of this product

Catalog No. AAL1120906



Specifications

Specifications

6-Fluorochroman-4-one
5g
C9H7FO2
4245390
SWBBIJZMIGAZHW-UHFFFAOYSA-N
6-fluoro-2,3-dihydrochromen-4-one
2733262
97%
114°C to 115°C
66892-34-0
MFCD00038654
6-fluorochroman-4-one, 6-fluoro-4-chromanone, 6-fluorochromanone, 6-fluoro-2,3-dihydro-4h-1-benzopyran-4-one, 6-fluorochromano-4-one, 6-fluoro-3,4-dihydro-2h-1-benzopyran-4-one, 6-fluoro-2,3-dihydro-4h-chromen-4-one, 6-fluoro-4-oxochroman, 6-fluoro-2,3-dihydro-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 6-fluoro-2,3-dihydro
C1COC2=C(C1=O)C=C(C=C2)F
166.151
166.15

Chemical Identifiers

66892-34-0
166.151
SWBBIJZMIGAZHW-UHFFFAOYSA-N
2733262
C1COC2=C(C1=O)C=C(C=C2)F
C9H7FO2
MFCD00038654
6-fluorochroman-4-one, 6-fluoro-4-chromanone, 6-fluorochromanone, 6-fluoro-2,3-dihydro-4h-1-benzopyran-4-one, 6-fluorochromano-4-one, 6-fluoro-3,4-dihydro-2h-1-benzopyran-4-one, 6-fluoro-2,3-dihydro-4h-chromen-4-one, 6-fluoro-4-oxochroman, 6-fluoro-2,3-dihydro-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 6-fluoro-2,3-dihydro
6-fluoro-2,3-dihydrochromen-4-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents