missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Fluoro-m-anisaldehyde, TCI America™

Manufacturer:  TCI America F038325G

 View more versions of this product

Catalog No. F038325G



Specifications

Specifications

6-Fluoro-m-anisaldehyde
White-Yellow
105728-90-3
MFCD00070795
DKIQXHIAEMGZGO-UHFFFAOYSA-N
2-fluoro-5-methoxybenzaldehyde
2734872
Crystal-Lump at 20°C
230°C
25g
C8H7FO2
6-fluoro-m-anisaldehyde, 2-fluoro-5-methoxy-benzaldehyde, 2-fluoro-5-methoxy benzaldehyde, benzaldehyde, 2-fluoro-5-methoxy, pubchem2634, 4-fluoro-3-formylanisole, acmc-2098hi, ksc490m2b, 2-fluoro-5-methyloxy benzaldehyde, 2-fluoro-5-methoxybenzaldehyde
COC1=CC(=C(C=C1)F)C=O
154.14
154.14
≥95.0% (GC)

Chemical Identifiers

105728-90-3
154.14
DKIQXHIAEMGZGO-UHFFFAOYSA-N
2734872
COC1=CC(=C(C=C1)F)C=O
C8H7FO2
MFCD00070795
6-fluoro-m-anisaldehyde, 2-fluoro-5-methoxy-benzaldehyde, 2-fluoro-5-methoxy benzaldehyde, benzaldehyde, 2-fluoro-5-methoxy, pubchem2634, 4-fluoro-3-formylanisole, acmc-2098hi, ksc490m2b, 2-fluoro-5-methyloxy benzaldehyde, 2-fluoro-5-methoxybenzaldehyde
2-fluoro-5-methoxybenzaldehyde
Safety and Handling

Safety and Handling

TSCA : No

Documents