Please login to your online account to display your discounted pricing

6-Fluoro-4H-1,3-benzodioxine-8-carboxylic acid, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC01901CB

 View more versions of this product

Catalog No. CC01901CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 198.15
Quantity 250mg
Assay 97%
CAS Max % 100.0
Molecular Formula C9H7FO4
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS Min % 96.5
MDL Number MFCD02681981
Chemical Name or Material 6-Fluoro-4H-1, 3-benzodioxine-8-carboxylic acid
Assay Percent Range 97%
CAS 321309-28-8

Specifications

Chemical Name or Material 6-Fluoro-4H-1, 3-benzodioxine-8-carboxylic acid
CAS 321309-28-8
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H7FO4
Formula Weight 198.15
MDL Number MFCD02681981
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents