Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Fluoro-4-hydroxy-2-methylquinoline, 97%

$87.84 - $325.46

Chemical Identifiers

Chemical Name or Material 6-Fluoro-4-hydroxy-2-methylquinoline
CAS 15912-68-2
Molecular Formula C10H8FNO
Formula Weight 177.18
MDL Number MFCD00041442
Beilstein 1526996
Synonym 6-Fluoro-4-hydroxyquinaldine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0761203

  SDS
alfa aesar™
L0761203
1g Each for $87.84

AAL0761206

  SDS
alfa aesar™
L0761206
5g Each for $325.46
Description & Specifications

Specifications

Formula Weight 177.18
CAS 15912-68-2
MDL Number MFCD00041442
Chemical Name or Material 6-Fluoro-4-hydroxy-2-methylquinoline
Synonym 6-Fluoro-4-hydroxyquinaldine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Quantity 1g
Health Hazard 1 Warning
Molecular Formula C10H8FNO
Beilstein 1526996
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈275°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.